Search
Search

Agxt| Gene Symbol: Agxt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276710.1 | NP_001263639.1 | 2 | 425 | ||
| NM_016702.3 | NP_057911.2 | 2 | 425 | |||
| GenBank mRNA | AF027730.2 | - | 2 | 420 | ||
| AK149432.1 | - | 2 | 348 | |||
| BC025799.1 | - | 2 | 389 | 5099627 | ||