Search
Search

Fabp4| Gene Symbol: Fabp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024406.2 | NP_077717.1 | 1 | 120 | ||
| GenBank mRNA | AK003143.1 | - | 1 | 121 | ||
| AK088535.1 | - | 1 | 121 | |||
| BC018558.1 | - | 1 | 102 | |||
| BC054426.1 | - | 1 | 103 | 5029359 | ||
| BY081693.1 | - | 1 | 120 | |||
| CT010390.1 | - | 1 | 55 | |||
| K02109.1 | - | 1 | 98 | |||
| M28726.1 | - | 1 | 25 | |||