Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BCL2 SIRNA Assays ›
          Gene Symbol:
          Bcl2
          Gene Name:
          B cell leukemia/lymphoma 2
          Gene Aliases:
          AW986256, Bcl-2, C430015F12Rik, D630044D05Rik, D830018M01Rik
          Chromosome Location:
          Chr.1: 106538178 - 106714290 on Build GRCm38
          UniGene ID:
          Mm.257460
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s62911

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Bcl2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_009741.4 NP_033871.2 1 1476
          NM_177410.2 NP_803129.2 1 1476
          GenBank mRNA AA867214.1 - 1 363
          AK049473.1 - 1 1470
          AK077913.1 - 1 256
          BC068988.1 - 1 364 1265430
          BC095964.1 - 1 1132 6306635

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: AF512835.

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          cell morphogenesis
          response to acid chemical
          ossification
          ovarian follicle development
          metanephros development
          ureteric bud development
          branching involved in ureteric bud morphogenesis
          behavioral fear response
          leukocyte homeostasis
          B cell homeostasis
          kidney development
          release of cytochrome c from mitochondria
          regulation of cell-matrix adhesion
          lymphocyte homeostasis
          lymphoid progenitor cell differentiation
          B cell lineage commitment
          T cell lineage commitment
          immune system development
          response to ischemia
          renal system process
          protein dephosphorylation
          melanin metabolic process
          regulation of nitrogen utilization
          cellular calcium ion homeostasis
          apoptotic process
          cellular response to DNA damage stimulus
          response to oxidative stress
          actin filament organization
          axonogenesis
          cell aging
          cell proliferation
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          male gonad development
          extrinsic apoptotic signaling pathway via death domain receptors
          intrinsic apoptotic signaling pathway in response to DNA damage
          intrinsic apoptotic signaling pathway in response to oxidative stress
          apoptotic mitochondrial changes
          response to toxic substance
          post-embryonic development
          organ morphogenesis
          response to iron ion
          response to UV-B
          response to gamma radiation
          regulation of gene expression
          negative regulation of autophagy
          negative regulation of calcium ion transport into cytosol
          regulation of glycoprotein biosynthetic process
          mesenchymal cell development
          positive regulation of neuron maturation
          positive regulation of smooth muscle cell migration
          cell growth
          single organismal cell-cell adhesion
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          cochlear nucleus development
          gland morphogenesis
          regulation of transmembrane transporter activity
          hemopoiesis
          B cell differentiation
          T cell differentiation
          negative regulation of ossification
          positive regulation of cell growth
          negative regulation of cell growth
          melanocyte differentiation
          negative regulation of cell migration
          positive regulation of B cell proliferation
          hair follicle morphogenesis
          axon regeneration
          regulation of protein stability
          endoplasmic reticulum calcium ion homeostasis
          glomerulus development
          negative regulation of cellular pH reduction
          regulation of protein localization
          negative regulation of myeloid cell apoptotic process
          T cell differentiation in thymus
          positive regulation of peptidyl-serine phosphorylation
          negative regulation of osteoblast proliferation
          response to cytokine
          response to nicotine
          organ growth
          growth
          positive regulation of multicellular organism growth
          B cell proliferation
          cellular response to glucose starvation
          response to drug
          response to hydrogen peroxide
          regulation of apoptotic process
          T cell homeostasis
          negative regulation of apoptotic process
          regulation of programmed cell death
          positive regulation of catalytic activity
          CD8-positive, alpha-beta T cell lineage commitment
          pigmentation
          regulation of protein homodimerization activity
          regulation of protein heterodimerization activity
          negative regulation of neuron apoptotic process
          ear development
          regulation of viral genome replication
          positive regulation of melanocyte differentiation
          negative regulation of mitotic cell cycle
          negative regulation of retinal cell programmed cell death
          regulation of mitochondrial membrane permeability
          focal adhesion assembly
          developmental pigmentation
          regulation of developmental pigmentation
          positive regulation of developmental pigmentation
          spleen development
          thymus development
          response to steroid hormone
          digestive tract morphogenesis
          developmental growth
          oocyte development
          positive regulation of skeletal muscle fiber development
          pigment granule organization
          homeostasis of number of cells within a tissue
          regulation of catalytic activity
          B cell receptor signaling pathway
          response to glucocorticoid
          neuron apoptotic process
          defense response to virus
          regulation of cell cycle
          regulation of mitochondrial membrane potential
          regulation of calcium ion transport
          transmembrane transport
          intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
          cellular response to organic substance
          cellular response to hypoxia
          reactive oxygen species metabolic process
          extrinsic apoptotic signaling pathway in absence of ligand
          negative regulation of G1/S transition of mitotic cell cycle
          negative regulation of reactive oxygen species metabolic process
          negative regulation of anoikis
          negative regulation of apoptotic signaling pathway
          negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
          negative regulation of intrinsic apoptotic signaling pathway
          protease binding
          protein binding
          transcription factor binding
          channel activity
          channel inhibitor activity
          protein phosphatase binding
          ubiquitin protein ligase binding
          identical protein binding
          protein homodimerization activity
          protein heterodimerization activity
          BH domain binding
          BH3 domain binding
          protein phosphatase 2A binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-qq54g:80/100.66.72.153:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0