Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BMP2 SIRNA Assays ›
          Gene Symbol:
          Bmp2
          Gene Name:
          bone morphogenetic protein 2
          Gene Aliases:
          AI467020, Bmp2a
          Chromosome Location:
          Chr.2: 133552159 - 133562896 on Build GRCm38
          UniGene ID:
          Mm.103205
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s63017

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Bmp2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_007553.3 NP_031579.2 3 1658
          GenBank mRNA AK132433.1 - 3 1623
          AK133923.1 - 3 1658
          AK161862.1 - 3 1462
          BC100344.1 - 3 618 30846365

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          activation of MAPK activity
          ossification
          osteoblast differentiation
          branching involved in ureteric bud morphogenesis
          response to hypoxia
          in utero embryonic development
          epithelial to mesenchymal transition
          positive regulation of protein phosphorylation
          positive regulation of endothelial cell proliferation
          chondrocyte differentiation
          BMP signaling pathway involved in heart induction
          atrioventricular valve morphogenesis
          endocardial cushion morphogenesis
          negative regulation of Wnt signaling pathway involved in heart development
          proteoglycan metabolic process
          regulation of transcription, DNA-templated
          protein phosphorylation
          transforming growth factor beta receptor signaling pathway
          Notch signaling pathway
          multicellular organism development
          heart development
          negative regulation of cell proliferation
          embryo development
          organ morphogenesis
          positive regulation of gene expression
          negative regulation of gene expression
          positive regulation of epithelial to mesenchymal transition
          positive regulation of pathway-restricted SMAD protein phosphorylation
          negative regulation of steroid biosynthetic process
          positive regulation of phosphatase activity
          telencephalon development
          telencephalon regionalization
          cell differentiation
          positive regulation of Wnt signaling pathway
          bone mineralization
          positive regulation of cell migration
          positive regulation of bone mineralization
          BMP signaling pathway
          protein destabilization
          negative regulation of aldosterone biosynthetic process
          positive regulation of osteoblast proliferation
          cardiocyte differentiation
          embryonic heart tube anterior/posterior pattern specification
          bone mineralization involved in bone maturation
          growth
          odontogenesis of dentin-containing tooth
          positive regulation of odontogenesis
          regulation of odontogenesis of dentin-containing tooth
          regulation of apoptotic process
          positive regulation of apoptotic process
          regulation of MAPK cascade
          positive regulation of MAPK cascade
          negative regulation of insulin-like growth factor receptor signaling pathway
          cell fate commitment
          positive regulation of cell differentiation
          positive regulation of fat cell differentiation
          positive regulation of neuron differentiation
          positive regulation of osteoblast differentiation
          positive regulation of ossification
          negative regulation of cell cycle
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          cell development
          positive regulation of astrocyte differentiation
          mesenchymal cell differentiation
          inner ear development
          positive regulation of neurogenesis
          negative regulation of calcium-independent cell-cell adhesion
          cartilage development
          cardiac muscle cell differentiation
          cardiac muscle tissue morphogenesis
          oxidation-reduction process
          pericardium development
          corticotropin hormone secreting cell differentiation
          thyroid-stimulating hormone-secreting cell differentiation
          cardiac epithelial to mesenchymal transition
          pathway-restricted SMAD protein phosphorylation
          SMAD protein signal transduction
          mesenchyme development
          positive regulation of Wnt signaling pathway by BMP signaling pathway
          positive regulation of cartilage development
          positive regulation of ERK1 and ERK2 cascade
          cellular response to growth factor stimulus
          cellular response to organic cyclic compound
          cellular response to BMP stimulus
          mesenchymal cell proliferation involved in ureteric bud development
          positive regulation of p38MAPK cascade
          positive regulation of transcription from RNA polymerase II promoter involved in cellular response to chemical stimulus
          negative regulation of cortisol biosynthetic process
          negative regulation of cardiac muscle cell differentiation
          retinol dehydrogenase activity
          receptor binding
          cytokine activity
          transforming growth factor beta receptor binding
          protein binding
          growth factor activity
          phosphatase activator activity
          protein domain specific binding
          protein homodimerization activity
          SMAD binding
          protein heterodimerization activity
          BMP receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0