Search Thermo Fisher Scientific
Gene Symbol: Brca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081001.2 | NP_001074470.1 | 4 | 816 | ||
NM_009765.3 | NP_033895.2 | 4 | 622 | |||
GenBank mRNA | AK145017.1 | - | 4 | 783 | ||
AK148272.1 | - | 4 | 620 | |||
AK164598.1 | - | 4 | 612 | |||
U65594.1 | - | 4 | 599 | |||
U72947.1 | - | 3 | 258 | |||
U82270.1 | - | 3 | 308 | |||
U89652.1 | - | 3 | 346 |