Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cbln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019626.3 | NP_062600.2 | 2 | 639 | ||
GenBank mRNA | AK158818.1 | - | 2 | 640 | ||
AK158885.1 | - | 2 | 657 | |||
AK161846.1 | - | 2 | 238 | |||
BC051956.1 | - | 2 | 254 | |||
BC055730.1 | - | 2 | 343 | 6404555 | ||
BC132120.1 | - | 3 | 322 | 40130397 | ||
BC132122.1 | - | 3 | 388 | 40130399 |