Search
Search

Cd33| Gene Symbol: Cd33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111058.1 | NP_001104528.1 | 2 | 380 | ||
| NM_021293.3 | NP_067268.1 | 2 | 380 | |||
| GenBank mRNA | AK046303.1 | - | 4 | 603 | ||
| AK089205.1 | - | 2 | 297 | |||
| AK155745.1 | - | 2 | 309 | |||
| AK170937.1 | - | 2 | 348 | |||
| BC132379.1 | - | 2 | 380 | 40130656 | ||
| S71345.1 | - | 2 | 311 | |||
| S71403.1 | - | 2 | 311 | |||