Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CITED1 SIRNA Assays ›
          Gene Symbol:
          Cited1
          Gene Name:
          Cbp/p300-interacting transactivator with Glu/Asp-rich carboxy-terminal domain 1
          Gene Aliases:
          AI316840, AU019144, Msg1
          Chromosome Location:
          Chr.X: 102247378 - 102252181 on Build GRCm38
          UniGene ID:
          Mm.2390
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s63999

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Cited1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001276466.1 NP_001263395.1 3 513
          NM_001276473.1 NP_001263402.1 3 382
          NM_001276474.1 NP_001263403.1 3 629
          NM_007709.4 NP_031735.1 3 716
          GenBank mRNA AK148042.1 - 3 495
          BC052030.1 - 3 642 5709775
          U65091.1 - 3 495

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_172055, XM_006257098, XM_006257097, XM_006257099, XM_006257100, and others.

          Panther Classification:

          Molecular Function -

          transcription cofactor

          Gene Ontology Categories:

          Function(s) Process(es)

          embryonic axis specification
          vasculogenesis
          branching involved in ureteric bud morphogenesis
          kidney development
          placenta development
          morphogenesis of an epithelium
          negative regulation of mesenchymal to epithelial transition involved in metanephros morphogenesis
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          nucleocytoplasmic transport
          apoptotic process
          transforming growth factor beta receptor signaling pathway
          multicellular organism development
          positive regulation of gene expression
          cell differentiation
          melanocyte differentiation
          response to lipopolysaccharide
          response to insulin
          response to cytokine
          response to interferon-gamma
          pigmentation
          negative regulation of neuron apoptotic process
          response to estrogen
          negative regulation of osteoblast differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          response to cAMP
          SMAD protein signal transduction
          labyrinthine layer development
          spongiotrophoblast layer development
          response to interleukin-1
          response to interleukin-2
          response to interleukin-4
          response to interleukin-6
          response to interleukin-9
          response to interleukin-11
          response to parathyroid hormone
          response to transforming growth factor beta
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          transcription coactivator activity
          protein binding
          protein C-terminus binding
          protein homodimerization activity
          transcription regulatory region DNA binding
          LBD domain binding
          co-SMAD binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0