Search
Search

Cnn2| Gene Symbol: Cnn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007725.2 | NP_031751.1 | 5 | 506 | ||
| GenBank mRNA | AK051845.1 | - | 5 | 507 | ||
| AK189304.1 | - | 5 | 434 | |||
| BC008181.1 | - | 5 | 424 | 3589470 | ||
| BC009144.1 | - | 5 | 461 | 3594856 | ||
| BC018482.1 | - | 5 | 476 | 3589158 | ||