Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
Unity Lab Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CRH SIRNA Assays ›
Gene Symbol:
Crh
Gene Name:
corticotropin releasing hormone
Gene Aliases:
CRF, Gm1347
Chromosome Location:
Chr.3: 19693401 - 19695396 on Build GRCm38
UniGene ID:
Mm.290689
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
s64389
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Crh
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_205769.2
NP_991338.1
2
1074
GenBank mRNA
BC116921.1
-
1
856
40126240
BC119036.1
-
1
856
8733785
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_031019, X03036.
Panther Classification:
Molecular Function -
peptide hormone
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of protein phosphorylation
synaptic transmission, dopaminergic
glucocorticoid biosynthetic process
inflammatory response
female pregnancy
learning or memory
positive regulation of cell proliferation
associative learning
hormone-mediated apoptotic signaling pathway
positive regulation of gene expression
negative regulation of gene expression
negative regulation of norepinephrine secretion
positive regulation of cell death
regulation of serotonin secretion
lung development
adrenal gland development
positive regulation of cAMP biosynthetic process
negative regulation of epinephrine secretion
negative regulation of luteinizing hormone secretion
locomotory exploration behavior
positive regulation of insulin secretion involved in cellular response to glucose stimulus
negative regulation of blood pressure
positive regulation of corticotropin secretion
positive regulation of cortisol secretion
long-term synaptic potentiation
positive regulation of digestive system process
negative regulation of cell death
negative regulation of glucagon secretion
cellular response to cocaine
positive regulation of calcium ion import
regulation of N-methyl-D-aspartate selective glutamate receptor activity
positive regulation of corticosterone secretion
positive regulation of behavioral fear response
hormone activity
corticotropin-releasing hormone receptor 1 binding
corticotropin-releasing hormone receptor 2 binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0