Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dclk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111051.1 | NP_001104521.1 | 4 | 574 | ||
NM_001111052.1 | NP_001104522.1 | 4 | 574 | |||
NM_001195538.1 | NP_001182467.1 | 8 | 1435 | |||
NM_001195539.1 | NP_001182468.1 | 4 | 622 | |||
NM_019978.3 | NP_064362.1 | 8 | 1483 | |||
GenBank mRNA | AB093234.1 | - | 8 | 1392 | ||
AF155819.1 | - | 8 | 1381 | |||
AF155820.1 | - | 8 | 1374 | |||
AF155821.1 | - | 4 | 433 | |||
AK032424.1 | - | 4 | 464 | |||
AK047716.1 | - | 4 | 361 | |||
AK081200.1 | - | 4 | 417 | |||
AK163437.1 | - | 2 | 792 | |||
AY968047.1 | - | 4 | 322 | |||
AY968048.1 | - | 4 | 325 | |||
BC133685.1 | - | 8 | 1237 | 40110683 |