Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DDX5 SIRNA Assays ›
          Gene Symbol:
          Ddx5
          Gene Name:
          DEAD (Asp-Glu-Ala-Asp) box polypeptide 5
          Gene Aliases:
          2600009A06Rik, G17P1, HUMP68, Hlr1, p68
          Chromosome Location:
          Chr.11: 106780355 - 106789268 on Build GRCm38
          UniGene ID:
          Mm.491096
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s64911

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 404.00
          Your Price
          Online offer: 391.65
          391.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ddx5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_007840.3 NP_031866.2 13 1911
          GenBank mRNA AK013106.1 - 6 800
          AK044792.1 - 13 1912
          AK076018.1 - 13 1921
          AK088887.1 - 13 1912
          AK129022.1 - 12 3185
          AK132779.1 - 13 1958
          AK135173.1 - 13 1912
          AK166045.1 - 13 1912
          AK166698.1 - 14 2044
          AK184106.1 - 2 351
          BC009142.1 - 2 1224
          BC046554.1 - 13 1889
          BC062916.1 - 13 1896 6827275
          BC086320.1 - 13 1834
          BC108369.1 - 13 1756 6312791
          BC129873.1 - 13 1789 40110366
          BC129874.1 - 13 1792 40110367
          X65627.1 - 13 1893

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001007613, XM_008768312, XM_008768313, FQ227563, BC079036.

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          regulation of alternative mRNA splicing, via spliceosome
          in utero embryonic development
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          mRNA processing
          circadian rhythm
          RNA splicing
          mRNA transcription
          RNA secondary structure unwinding
          positive regulation of intracellular estrogen receptor signaling pathway
          positive regulation of DNA damage response, signal transduction by p53 class mediator
          regulation of viral genome replication
          regulation of osteoblast differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          rhythmic process
          regulation of androgen receptor signaling pathway
          intrinsic apoptotic signaling pathway by p53 class mediator
          regulation of skeletal muscle cell differentiation
          nucleotide binding
          nucleic acid binding
          transcription cofactor activity
          transcription coactivator activity
          RNA binding
          RNA helicase activity
          ATP-dependent RNA helicase activity
          helicase activity
          protein binding
          calmodulin binding
          ATP binding
          hydrolase activity
          hydrolase activity, acting on acid anhydrides, in phosphorus-containing anhydrides
          enzyme binding
          estrogen receptor binding
          pre-mRNA binding
          poly(A) RNA binding
          calcium-dependent protein binding
          androgen receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0