Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: Ebf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276387.1 | NP_001263316.1 | 3 | 1131 | ||
NM_010095.6 | NP_034225.1 | 3 | 1126 | |||
GenBank mRNA | AK053013.1 | - | 3 | 1127 | ||
AK139262.1 | - | 3 | 838 | |||
BC049188.1 | - | 2 | 425 | 6312260 | ||
BC050922.1 | - | 2 | 648 | 6409737 | ||
U71189.1 | - | 2 | 311 | |||
U82441.1 | - | 2 | 180 | |||
U92703.1 | - | 2 | 531 |