Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EEF1A1 SIRNA Assays ›
Gene Symbol:
Eef1a1
Gm6548
Gene Name:
eukaryotic translation elongation factor 1 alpha 1
eukaryotic translation elongation factor 1 alpha 1 pseudogene
Gene Aliases:
-
EG625054
Chromosome Location:
Chr.9: 78478453 - 78481724 on Build GRCm38
Chromosome Location:
Chr.17: 78850504 - 78852544 on Build GRCm38
UniGene ID:
Mm.138471
Mm.311918
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID
s65322
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Eef1a1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_010106.2
NP_034236.2
4
572
GenBank mRNA
AK011771.1
-
4
520
AK012341.1
-
4
521
AK032914.1
-
4
518
AK050635.1
-
4
519
AK076696.1
-
4
520
AK081725.1
-
4
520
AK083361.1
-
4
520
AK088041.1
-
4
521
AK133725.1
-
4
520
AK135448.1
-
4
521
AK137223.1
-
4
520
AK141243.1
-
4
522
AK150798.1
-
4
520
AK151479.1
-
4
521
AK151773.1
-
4
520
AK152810.1
-
4
520
AK152993.1
-
4
521
AK153387.1
-
4
520
AK153402.1
-
4
520
AK153851.1
-
4
520
AK162049.1
-
4
520
AK162313.1
-
4
519
AK162440.1
-
4
524
AK163799.1
-
4
520
AK164113.1
-
4
522
AK164671.1
-
4
520
AK165938.1
-
4
520
AK166021.1
-
4
520
AK166033.1
-
4
520
AK166058.1
-
4
520
AK166089.1
-
4
520
AK166615.1
-
4
520
AK166807.1
-
4
520
AK167843.1
-
4
520
AK167873.1
-
4
519
AK168424.1
-
4
520
AK168467.1
-
4
526
AK187140.1
-
1
53
AK188863.1
-
2
213
AK206299.1
-
1
53
AK209036.1
-
1
51
AK210348.1
-
1
54
AK213893.1
-
1
53
BC003969.1
-
3
490
BC004005.1
-
3
481
3492770
BC004067.1
-
3
509
3590965
BC005660.1
-
3
510
3588966
BC018223.1
-
4
508
4012653
BC018485.1
-
4
522
3491827
BC040689.1
-
1
12
BC050124.1
-
2065
BC068258.1
-
2317
BC083069.1
-
4
508
6310448
BC092053.1
-
3
498
4008574
BC092276.1
-
3
503
3490007
BC092292.1
-
3
464
BC095965.1
-
3
498
6310404
BC108391.1
-
4
505
30242277
M22432.1
-
4
511
X13661.1
-
3
491
Gene Symbol: Gm6548
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
GenBank mRNA
BC089461.1
-
1
818
Back To Top
More Information
Panther Classification:
Molecular Function -
translation factor
Gene Ontology Categories:
Function(s)
Process(es)
translation
translational elongation
protein homooligomerization
cellular response to epidermal growth factor stimulus
tRNA binding
nucleotide binding
mRNA binding
translation elongation factor activity
GTPase activity
protein binding
GTP binding
protein kinase binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0