Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other EEF1A1 SIRNA Assays ›
          Gene Symbol:
          Eef1a1 Gm6548
          Gene Name:
          eukaryotic translation elongation factor 1 alpha 1
          eukaryotic translation elongation factor 1 alpha 1 pseudogene
          Gene Aliases:
          -
          EG625054
          Chromosome Location:
          Chr.9: 78478453 - 78481724 on Build GRCm38
          Chromosome Location:
          Chr.17: 78850504 - 78852544 on Build GRCm38
          UniGene ID:
          Mm.138471
          Mm.311918
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s65323

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Eef1a1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_010106.2 NP_034236.2 6 1066
          GenBank mRNA AK003188.1 - 6 1003
          AK011771.1 - 6 1015
          AK012341.1 - 6 1015
          AK032914.1 - 6 1012
          AK050635.1 - 6 1015
          AK076696.1 - 6 1014
          AK081725.1 - 6 1014
          AK083361.1 - 6 1014
          AK088041.1 - 6 1015
          AK133725.1 - 6 1014
          AK135448.1 - 6 1015
          AK137223.1 - 6 1014
          AK141243.1 - 6 1016
          AK150798.1 - 6 1014
          AK151479.1 - 6 1015
          AK151773.1 - 6 1014
          AK152810.1 - 6 1014
          AK152993.1 - 6 1015
          AK153387.1 - 6 1014
          AK153402.1 - 6 1014
          AK153851.1 - 6 1016
          AK162049.1 - 6 1014
          AK162313.1 - 6 1013
          AK162440.1 - 6 1018
          AK163799.1 - 6 1014
          AK164113.1 - 6 1016
          AK164671.1 - 6 1014
          AK165938.1 - 6 1014
          AK166021.1 - 6 1014
          AK166033.1 - 6 1014
          AK166058.1 - 6 1014
          AK166089.1 - 6 1014
          AK166615.1 - 6 1014
          AK166807.1 - 6 1014
          AK167843.1 - 6 1014
          AK167873.1 - 6 1013
          AK168424.1 - 6 1016
          AK168467.1 - 6 1022
          BC003969.1 - 5 984
          BC004005.1 - 5 975 3492770
          BC004067.1 - 5 1003 3590965
          BC005660.1 - 5 1004 3588966
          BC018223.1 - 6 1002 4012653
          BC018485.1 - 6 1016 3491827
          BC040689.1 - 3 506
          BC050124.1 - 2559
          BC068258.1 - 2811
          BC083069.1 - 6 1002 6310448
          BC092053.1 - 5 992 4008574
          BC092276.1 - 5 997 3490007
          BC092292.1 - 5 957
          BC095965.1 - 5 992 6310404
          BC108391.1 - 6 999 30242277
          M22432.1 - 6 1005
          X13661.1 - 5 982
          Gene Symbol: Gm6548
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC089461.1 - 1 1313

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          translation factor

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          translational elongation
          protein homooligomerization
          cellular response to epidermal growth factor stimulus
          tRNA binding
          nucleotide binding
          mRNA binding
          translation elongation factor activity
          GTPase activity
          protein binding
          GTP binding
          protein kinase binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0