Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EEF1A1 SIRNA Assays ›
Gene Symbol:
Eef1a1
Gene Name:
eukaryotic translation elongation factor 1 alpha 1
Gene Aliases:
-
Chromosome Location:
Chr.9: 78478453 - 78481724 on Build GRCm38
UniGene ID:
Mm.138471
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
s65324
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Eef1a1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_010106.2
NP_034236.2
4
629
GenBank mRNA
AK003188.1
-
4
575
AK011771.1
-
4
577
AK012341.1
-
4
577
AK032914.1
-
4
575
AK050635.1
-
4
576
AK076696.1
-
4
577
AK081725.1
-
4
577
AK083361.1
-
4
577
AK088041.1
-
4
578
AK133725.1
-
4
577
AK135448.1
-
4
578
AK137223.1
-
4
577
AK141243.1
-
4
579
AK150798.1
-
4
577
AK151479.1
-
4
578
AK151773.1
-
4
577
AK152810.1
-
4
577
AK152993.1
-
4
578
AK153387.1
-
4
577
AK153402.1
-
4
577
AK153851.1
-
4
577
AK162049.1
-
4
577
AK162313.1
-
4
576
AK162440.1
-
4
581
AK163799.1
-
4
577
AK164113.1
-
4
579
AK164671.1
-
4
577
AK165938.1
-
4
577
AK166021.1
-
4
577
AK166033.1
-
4
577
AK166058.1
-
4
577
AK166089.1
-
4
577
AK166615.1
-
4
577
AK166807.1
-
4
577
AK167843.1
-
4
577
AK167873.1
-
4
576
AK168424.1
-
4
577
AK168467.1
-
4
583
AK187140.1
-
1
110
AK188863.1
-
2
271
AK206299.1
-
1
110
AK209036.1
-
1
108
AK210348.1
-
1
111
AK213893.1
-
1
110
BC003969.1
-
3
547
BC004005.1
-
3
538
3492770
BC004067.1
-
3
566
3590965
BC005660.1
-
3
567
3588966
BC018223.1
-
4
565
4012653
BC018485.1
-
4
579
3491827
BC040689.1
-
1
69
BC050124.1
-
2122
BC068258.1
-
2374
BC083069.1
-
4
565
6310448
BC092053.1
-
3
555
4008574
BC092276.1
-
3
560
3490007
BC092292.1
-
3
521
BC095965.1
-
3
555
6310404
BC108391.1
-
4
562
30242277
M22432.1
-
4
568
X13661.1
-
3
548
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_175838, BC111707, L10339, X61043, X63561, and others.
Panther Classification:
Molecular Function -
translation factor
Gene Ontology Categories:
Function(s)
Process(es)
translation
translational elongation
protein homooligomerization
cellular response to epidermal growth factor stimulus
tRNA binding
nucleotide binding
mRNA binding
translation elongation factor activity
GTPase activity
protein binding
GTP binding
protein kinase binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-9c8cf:80/100.66.75.150:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0