Search
Search

Eif1a| Gene Symbol: Eif1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010120.5 | NP_034250.3 | 3 | 743 | ||
| GenBank mRNA | AF026481.1 | - | 2 | 1562 | ||
| AK009026.1 | - | 3 | 714 | |||
| AK013268.1 | - | 3 | 742 | |||
| AK019451.1 | - | 3 | 744 | |||
| AK150651.1 | - | 3 | 740 | |||
| AK150673.1 | - | 3 | 716 | |||
| AK150677.1 | - | 3 | 716 | |||
| AK166368.1 | - | 715 | ||||
| AK167333.1 | - | 3 | 703 | |||
| BC027437.1 | - | 3 | 705 | 4017054 | ||
| CT010335.1 | - | 1 | 390 | |||