Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ENG SIRNA Assays ›
          Gene Symbol:
          Eng
          Gene Name:
          endoglin
          Gene Aliases:
          AI528660, AI662476, CD105, Endo, S-endoglin
          Chromosome Location:
          Chr.2: 32646532 - 32682669 on Build GRCm38
          UniGene ID:
          Mm.225297
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s65507

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Eng
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001146348.1 NP_001139820.1 6 1147
          NM_001146350.1 NP_001139822.1 6 1150
          NM_007932.2 NP_031958.2 6 1150
          GenBank mRNA AK151302.1 - 6 1120
          AY679531.1 - 6 1150
          BC029080.1 - 6 1064 4168250
          S69407.1 - 7 1015
          X77952.1 - 6 1150

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          angiogenesis
          patterning of blood vessels
          vasculogenesis
          response to hypoxia
          positive regulation of protein phosphorylation
          negative regulation of endothelial cell proliferation
          heart looping
          positive regulation of systemic arterial blood pressure
          cell migration involved in endocardial cushion formation
          regulation of transcription, DNA-templated
          protein phosphorylation
          cell adhesion
          transmembrane receptor protein serine/threonine kinase signaling pathway
          transforming growth factor beta receptor signaling pathway
          heart development
          positive regulation of gene expression
          positive regulation of pathway-restricted SMAD protein phosphorylation
          cell migration
          regulation of transforming growth factor beta receptor signaling pathway
          central nervous system vasculogenesis
          extracellular matrix disassembly
          negative regulation of cell migration
          positive regulation of BMP signaling pathway
          negative regulation of protein autophosphorylation
          positive regulation of collagen biosynthetic process
          intracellular signal transduction
          wound healing
          positive regulation of angiogenesis
          positive regulation of transcription from RNA polymerase II promoter
          smooth muscle tissue development
          artery morphogenesis
          venous blood vessel morphogenesis
          cell motility
          negative regulation of nitric-oxide synthase activity
          cell chemotaxis
          negative regulation of pathway-restricted SMAD protein phosphorylation
          extracellular matrix constituent secretion
          detection of hypoxia
          transforming growth factor beta-activated receptor activity
          transforming growth factor beta receptor, cytoplasmic mediator activity
          type II transforming growth factor beta receptor binding
          protein binding
          galactose binding
          glycosaminoglycan binding
          type I transforming growth factor beta receptor binding
          BMP binding
          protein homodimerization activity
          transforming growth factor beta binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0