Search
Search

Ext2| Gene Symbol: Ext2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010163.3 | NP_034293.2 | 8 | 1449 | ||
| GenBank mRNA | AK082572.1 | - | 8 | 1431 | ||
| AK134438.1 | - | 4 | 390 | |||
| AK148090.1 | - | 8 | 1439 | |||
| AK148369.1 | - | 7 | 1210 | |||
| AK150155.1 | - | 8 | 1441 | |||
| AK152265.1 | - | 1549 | ||||
| AK152840.1 | - | 1551 | ||||
| AK164342.1 | - | 8 | 1435 | |||
| AK187641.1 | - | 3 | 252 | |||
| AK194704.1 | - | 3 | 299 | |||
| BC006597.1 | - | 8 | 1422 | 3965079 | ||
| U67837.1 | - | 8 | 1424 | |||
| U72141.1 | - | 8 | 1428 | |||