Search
Search

F8| Gene Symbol: F8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161373.1 | NP_001154845.1 | 8, 9 | 1677 | ||
| NM_001161374.1 | NP_001154846.1 | 8, 9 | 1677 | |||
| NM_007977.2 | NP_032003.2 | 8, 9 | 1677 | |||
| GenBank mRNA | BC138347.1 | - | 8, 9 | 1558 | 8861367 | |
| BC145245.1 | - | 8, 9 | 1558 | 9053797 | ||
| L05573.1 | - | 8, 9 | 1675 | |||