Search
Search

Fbp2| Gene Symbol: Fbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007994.3 | NP_032020.2 | 5 | 759 | ||
| GenBank mRNA | AJ132692.1 | - | 5 | 686 | ||
| AK146946.1 | - | 5 | 812 | |||
| AK166898.1 | - | 5 | 760 | |||
| AK166973.1 | - | 5 | 765 | |||
| AK168374.1 | - | 5 | 790 | |||
| BC012720.1 | - | 5 | 764 | 4205515 | ||
| D42083.1 | - | 5 | 705 | |||