Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FN1 SIRNA Assays ›
          Gene Symbol:
          Fn1
          Gene Name:
          fibronectin 1
          Gene Aliases:
          E330027I09, Fn, Fn-1
          Chromosome Location:
          Chr.1: 71585473 - 71653234 on Build GRCm38
          UniGene ID:
          Mm.193099
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s66182

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Fn1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001276408.1 NP_001263337.1 18 2929
          NM_001276409.1 NP_001263338.1 18 2929
          NM_001276410.1 NP_001263339.1 18 2929
          NM_001276411.1 NP_001263340.1 18 2929
          NM_001276412.1 NP_001263341.1 18 2929
          NM_001276413.1 NP_001263342.1 18 2929
          NM_010233.2 NP_034363.1 18 2929
          GenBank mRNA AK090130.1 - 18 2874
          AK090135.1 - 18 2870
          AK147249.1 - 9 1279
          AK147315.1 - 18 2868
          AK147639.1 - 18 2868
          AK147654.1 - 18 2868
          AK147663.1 - 18 2868
          AK147683.1 - 18 2868
          AK147701.1 - 18 2867
          AK170654.1 - 18 2868
          AK170755.1 - 18 2897
          AK171339.1 - 18 2868
          BC051082.1 - 18 2838
          BC138421.1 - 18 2793 8861441
          BC145271.1 - 18 2793 9053825
          CN538894.1 - 4 405

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          extracellular matrix structural protein

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          cell activation
          acute-phase response
          cell-substrate junction assembly
          cell adhesion
          cell-matrix adhesion
          calcium-independent cell-matrix adhesion
          glial cell migration
          regulation of cell shape
          positive regulation of peptidase activity
          peptide cross-linking
          extracellular matrix organization
          positive regulation of cell migration
          integrin activation
          substrate adhesion-dependent cell spreading
          endodermal cell differentiation
          wound healing
          negative regulation of apoptotic process
          positive regulation of axon extension
          positive regulation of chemotaxis
          cellular response to mercury ion
          cellular response to interleukin-1
          protease binding
          integrin binding
          protein binding
          heparin binding
          peptidase activator activity
          mercury ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0