Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GJA1 SIRNA Assays ›
          Gene Symbol:
          Gja1
          Gene Name:
          gap junction protein, alpha 1
          Gene Aliases:
          AU042049, AW546267, Cnx43, Cx43, Cx43alpha1, Gja-1, Npm1, connexin43
          Chromosome Location:
          Chr.10: 56377201 - 56390423 on Build GRCm38
          UniGene ID:
          Mm.378921
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s66668

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Gja1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_010288.3 NP_034418.1 2 1278
          GenBank mRNA AK029291.1 - 2 1135
          AK036979.1 - 2 1249
          AK081668.1 - 1 225
          AK145692.1 - 2 1248
          AK165986.1 - 2 1135
          AK171955.1 - 2 1246
          BC006894.1 - 2 1231 3599347
          BC011324.1 - 2 1360
          BC055375.1 - 2 1201 30063597
          CT010327.1 - 1 1046
          M63801.1 - 2 1266
          X61576.1 - 2 1132

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          gap junction

          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          in utero embryonic development
          neuron migration
          negative regulation of endothelial cell proliferation
          heart looping
          regulation of heart rate
          epithelial cell maturation
          lens development in camera-type eye
          chronic inflammatory response
          positive regulation of glomerular filtration
          atrial ventricular junction remodeling
          epicardial cell to mesenchymal cell transition
          apoptotic process
          cell communication
          signal transduction
          positive regulation of cytosolic calcium ion concentration
          cell-cell signaling
          heart development
          adult heart development
          regulation of heart contraction
          negative regulation of cell proliferation
          response to pH
          vascular transport
          positive regulation of gene expression
          negative regulation of gene expression
          cell communication by chemical coupling
          cell communication by electrical coupling
          positive regulation of cell communication by chemical coupling
          ATP transport
          regulation of bone mineralization
          positive regulation of insulin secretion
          ion transmembrane transport
          embryonic heart tube development
          embryonic digit morphogenesis
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          skeletal muscle tissue regeneration
          cell-cell junction organization
          positive regulation of osteoblast differentiation
          positive regulation of protein catabolic process
          positive regulation of striated muscle tissue development
          positive regulation of vasoconstriction
          positive regulation of vasodilation
          regulation of bone remodeling
          blood vessel morphogenesis
          neuron projection morphogenesis
          protein oligomerization
          regulation of calcium ion transport
          transmembrane transport
          negative regulation of cardiac muscle cell proliferation
          milk ejection
          regulation of ventricular cardiac muscle cell membrane repolarization
          regulation of blood vessel remodeling
          regulation of atrial cardiac muscle cell membrane depolarization
          regulation of ventricular cardiac muscle cell membrane depolarization
          negative regulation of wound healing
          cardiac conduction
          establishment of epithelial cell polarity
          negative regulation of DNA biosynthetic process
          regulation of bicellular tight junction assembly
          positive regulation of behavioral fear response
          signal transducer activity
          receptor binding
          gap junction channel activity
          protein binding
          ion transmembrane transporter activity
          SH3 domain binding
          protein domain specific binding
          transmembrane transporter activity
          PDZ domain binding
          beta-tubulin binding
          connexin binding
          scaffold protein binding
          gap junction channel activity involved in cell communication by electrical coupling

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-hvptj:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0