Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GLI3 SIRNA Assays ›
          Gene Symbol:
          Gli3
          Gene Name:
          GLI-Kruppel family member GLI3
          Gene Aliases:
          AI854843, AU023367, Bph, GLI3-190, GLI3FL, Pdn, Xt, add
          Chromosome Location:
          Chr.13: 15463235 - 15730026 on Build GRCm38
          UniGene ID:
          Mm.5098
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s66730

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Gli3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_008130.2 NP_032156.2 3 603
          GenBank mRNA AK144851.1 - 3 387
          AK158261.1 - 3 603
          BC141135.1 - 3 304 9055747
          BC145445.1 - 3 304 9054003
          X95255.1 - 3 343

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_000168, BC113616, BF194991, AK308429, BC117168, and others.
          This siRNA also targets the following transcripts in Rat: AB073718.

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          metanephros development
          branching involved in ureteric bud morphogenesis
          in utero embryonic development
          kidney development
          positive regulation of neuroblast proliferation
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          smoothened signaling pathway
          pattern specification process
          axon guidance
          central nervous system development
          brain development
          hindgut morphogenesis
          heart development
          negative regulation of cell proliferation
          anterior/posterior pattern specification
          dorsal/ventral pattern formation
          proximal/distal pattern formation
          regulation of gene expression
          protein processing
          spinal cord dorsal/ventral patterning
          spinal cord motor neuron differentiation
          telencephalon development
          pallium development
          subpallium development
          optic nerve morphogenesis
          hippocampus development
          smoothened signaling pathway involved in ventral spinal cord interneuron specification
          smoothened signaling pathway involved in spinal cord motor neuron cell fate specification
          forebrain dorsal/ventral pattern formation
          cerebral cortex radial glia guided migration
          layer formation in cerebral cortex
          forebrain radial glial cell differentiation
          neural tube development
          lateral ganglionic eminence cell proliferation
          melanocyte differentiation
          lung development
          embryonic limb morphogenesis
          mammary gland development
          forebrain development
          positive regulation of chondrocyte differentiation
          T cell differentiation in thymus
          limb morphogenesis
          tube development
          regulation of cell proliferation
          positive regulation of protein import into nucleus
          odontogenesis of dentin-containing tooth
          embryonic digit morphogenesis
          regulation of apoptotic process
          camera-type eye development
          negative regulation of apoptotic process
          tongue development
          negative thymic T cell selection
          regulation of cell differentiation
          negative regulation of cell differentiation
          negative regulation of neuron differentiation
          positive regulation of osteoblast differentiation
          negative regulation of smoothened signaling pathway
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of alpha-beta T cell differentiation
          negative regulation of alpha-beta T cell differentiation
          embryonic digestive tract morphogenesis
          embryonic digestive tract development
          developmental growth
          camera-type eye morphogenesis
          embryonic morphogenesis
          anatomical structure formation involved in morphogenesis
          neuron fate commitment
          embryonic skeletal system morphogenesis
          oligodendrocyte differentiation
          branching morphogenesis of an epithelial tube
          inner ear development
          anatomical structure development
          palate development
          limb development
          frontal suture morphogenesis
          lambdoid suture morphogenesis
          sagittal suture morphogenesis
          mammary gland specification
          smoothened signaling pathway involved in dorsal/ventral neural tube patterning
          artery development
          anterior semicircular canal development
          lateral semicircular canal development
          cell differentiation involved in kidney development
          thymocyte apoptotic process
          negative regulation of canonical Wnt signaling pathway
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          nucleic acid binding
          DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          beta-catenin binding
          histone acetyltransferase binding
          histone deacetylase binding
          sequence-specific DNA binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0