Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 企業服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HNF4A SIRNA Assays ›
          Gene Symbol:
          HNF4A
          Gene Name:
          hepatocyte nuclear factor 4 alpha
          Gene Aliases:
          FRTS4, HNF4, HNF4a7, HNF4a8, HNF4a9, HNF4alpha, MODY, MODY1, NR2A1, NR2A21, TCF, TCF14
          Chromosome Location:
          Chr.20: 44355801 - 44432845 on Build GRCh38
          UniGene ID:
          Hs.116462
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s6698

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HNF4A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000457.4 NP_000448.3 4 533
          NM_001030003.2 NP_001025174.1 4 354
          NM_001030004.2 NP_001025175.1 4 354
          NM_001258355.1 NP_001245284.1 5 624
          NM_001287182.1 NP_001274111.1 5 576
          NM_001287183.1 NP_001274112.1 5 576
          NM_001287184.1 NP_001274113.1 5 576
          NM_175914.4 NP_787110.2 4 354
          NM_178849.2 NP_849180.1 4 533
          NM_178850.2 NP_849181.1 4 533
          XM_005260407.3 XP_005260464.1 5 653
          GenBank mRNA AB307703.1 - 4 389
          AY680696.1 - 4 354
          AY680697.1 - 4 354
          AY680698.1 - 4 354
          BC137539.1 - 4 455 9021539
          BC137540.1 - 4 455 9021540
          BP274413.1 - 4 533
          HQ692860.1 - 4 416
          HQ692868.1 - 4 350
          HQ692869.1 - 4 416
          X76930.1 - 4 408
          X87870.1 - 4 488
          X87871.1 - 4 473
          X87872.1 - 4 509
          Z49825.1 - 5 609

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          C4 zinc finger nuclear receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription from RNA polymerase II promoter
          transcription initiation from RNA polymerase II promoter
          ornithine metabolic process
          lipid metabolic process
          acyl-CoA metabolic process
          xenobiotic metabolic process
          establishment of tissue polarity
          sex differentiation
          blood coagulation
          negative regulation of cell proliferation
          response to glucose
          regulation of gastrulation
          regulation of lipid metabolic process
          signal transduction involved in regulation of gene expression
          cell differentiation
          negative regulation of cell growth
          intracellular receptor signaling pathway
          regulation of microvillus assembly
          negative regulation of protein import into nucleus, translocation
          response to drug
          negative regulation of tyrosine phosphorylation of Stat5 protein
          glucose homeostasis
          steroid hormone mediated signaling pathway
          negative regulation of sequence-specific DNA binding transcription factor activity
          cell-cell junction organization
          positive regulation of gluconeogenesis
          positive regulation of fatty acid biosynthetic process
          positive regulation of transcription, DNA-templated
          negative regulation of mitotic cell cycle
          positive regulation of transcription from RNA polymerase II promoter
          regulation of insulin secretion
          response to cAMP
          lipid homeostasis
          phospholipid homeostasis
          SMAD protein signal transduction
          triglyceride homeostasis
          negative regulation of activation of JAK2 kinase activity
          positive regulation of cholesterol homeostasis
          fatty-acyl-CoA binding
          RNA polymerase II core promoter sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II activating transcription factor binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
          steroid hormone receptor activity
          RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
          receptor binding
          fatty acid binding
          protein binding
          zinc ion binding
          palmitoyl-CoA hydrolase activity
          protein homodimerization activity
          transcription regulatory region DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0