Search
Search

HNRNPF| Gene Symbol: HNRNPF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098204.1 | NP_001091674.1 | 4 | 1271 | ||
| NM_001098205.1 | NP_001091675.1 | 4 | 1311 | |||
| NM_001098206.1 | NP_001091676.1 | 4 | 1288 | |||
| NM_001098207.1 | NP_001091677.1 | 4 | 1315 | |||
| NM_001098208.1 | NP_001091678.1 | 3 | 1394 | |||
| NM_004966.3 | NP_004957.1 | 4 | 1272 | |||
| GenBank mRNA | AK001364.1 | - | 4 | 1309 | ||
| AK296696.1 | - | 5 | 1092 | |||
| BC001432.2 | - | 3 | 1066 | 3138435 | ||
| BC004254.1 | - | 4 | 1230 | 3633625 | ||
| BC015580.2 | - | 4 | 1187 | 4636807 | ||
| BC016736.1 | - | 4 | 1260 | 4104978 | ||
| BC106008.1 | - | 4 | 1199 | 6156823 | ||
| L28010.1 | - | 1 | 935 | |||