Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HDAC5 SIRNA Assays ›
          Gene Symbol:
          Hdac5
          Gene Name:
          histone deacetylase 5
          Gene Aliases:
          AI426555, Hdac4, mHDA1, mKIAA0600
          Chromosome Location:
          Chr.11: 102195747 - 102230186 on Build GRCm38
          UniGene ID:
          Mm.22665
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s67424

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 404.00
          Your Price
          Online offer: 391.65
          391.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Hdac5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001077696.1 NP_001071164.1 8 1148
          NM_001284248.1 NP_001271177.1 10 1660
          NM_001284249.1 NP_001271178.1 9 1257
          NM_001284250.1 NP_001271179.1 8 1145
          NM_010412.3 NP_034542.3 8 1145
          GenBank mRNA AK122325.2 - 3 184
          AK139202.1 - 9 1231
          AK145972.1 - 8 1111
          AK146483.1 - 8 1084
          AK155335.1 - 8 1119
          AK156008.1 - 5 1396
          AK157159.1 - 5 1396
          BC049926.1 - 8 1081
          BC058554.1 - 10 1612
          BC060609.1 - 7 939 5720438

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          histone modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          osteoblast development
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          protein deacetylation
          inflammatory response
          nervous system development
          heart development
          negative regulation of gene expression
          regulation of myotube differentiation
          negative regulation of myotube differentiation
          chromatin modification
          histone deacetylation
          B cell differentiation
          multicellular organismal response to stress
          regulation of gene expression, epigenetic
          B cell activation
          response to cocaine
          response to drug
          regulation of protein binding
          negative regulation of osteoblast differentiation
          negative regulation of striated muscle tissue development
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          regulation of skeletal muscle fiber development
          positive regulation of sequence-specific DNA binding transcription factor activity
          regulation of striated muscle cell differentiation
          renal tubule morphogenesis
          cellular response to fluid shear stress
          negative regulation of cell migration involved in sprouting angiogenesis
          positive regulation of neural precursor cell proliferation
          positive regulation of stem cell proliferation
          RNA polymerase III transcription factor binding
          core promoter sequence-specific DNA binding
          core promoter binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II transcription factor binding
          chromatin binding
          transcription corepressor activity
          histone deacetylase activity
          protein kinase C binding
          protein binding
          transcription factor binding
          hydrolase activity
          protein kinase binding
          histone deacetylase activity (H3-K14 specific)
          NAD-dependent histone deacetylase activity (H3-K14 specific)
          histone deacetylase activity (H3-K9 specific)
          protein deacetylase activity
          histone deacetylase activity (H4-K16 specific)
          histone deacetylase binding
          transcription regulatory region DNA binding
          metal ion binding
          NAD-dependent histone deacetylase activity (H3-K9 specific)
          NAD-dependent histone deacetylase activity (H4-K16 specific)
          repressing transcription factor binding
          NAD-dependent histone deacetylase activity (H3-K18 specific)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0