Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdgf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008231.4 | NP_032257.3 | 4 | 710 | ||
GenBank mRNA | AF251787.1 | - | 4 | 623 | ||
AK005359.1 | - | 4 | 489 | |||
AK017863.1 | - | 4 | 664 | |||
AK029475.1 | - | 4 | 454 | |||
AK076021.1 | - | 4 | 498 | |||
BC005713.1 | - | 4 | 620 | 2609261 | ||
BC021654.1 | - | 4 | 638 | 4163897 | ||
D63707.1 | - | 4 | 464 |