Search Thermo Fisher Scientific
Gene Symbol: Hspg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008305.3 | NP_032331.2 | 80 | 11106 | ||
GenBank mRNA | BC036291.1 | - | 6 | 722 | ||
BC085618.1 | - | 8 | 1198 | |||
BC094353.1 | - | 4 | 401 | |||
CD349187.1 | - | 2 | 211 |