Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Il17a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010552.3 | NP_034682.1 | 2 | 272 | ||
GenBank mRNA | AK040420.1 | - | 2 | 273 | ||
BC119303.1 | - | 2 | 272 | 8734052 | ||
BC119309.1 | - | 2 | 272 | 8734058 | ||
U43088.1 | - | 2 | 259 |