Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Insr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010568.2 | NP_034698.2 | 12 | 3103 | ||
GenBank mRNA | J05149.1 | - | 12 | 2662 | ||
KC356871.1 | - | 13 | 2650 |