Search
Search

Itgb1| Gene Symbol: Itgb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010578.2 | NP_034708.1 | 3 | 419 | ||
| GenBank mRNA | AK088016.1 | - | 3 | 320 | ||
| AK088729.1 | - | 3 | 166 | |||
| AK135521.1 | - | 3 | 318 | |||
| AK159689.1 | - | 3 | 261 | |||
| AK167682.1 | - | 3 | 317 | |||
| BC050906.1 | - | 3 | 292 | 5721348 | ||
| X15202.1 | - | 2 | 112 | |||
| Y00769.1 | - | 3 | 207 | |||