Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnj10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039484.1 | NP_001034573.1 | 2 | 1279 | ||
GenBank mRNA | AB039879.1 | - | 2 | 1136 | ||
AF322631.1 | - | 2 | 1088 | |||
AK044640.1 | - | 2 | 1203 | |||
AK048864.1 | - | 2 | 1282 | |||
AY374423.1 | - | 1 | 1046 | |||
BC099932.1 | - | 2 | 1222 | |||
BC141088.1 | - | 2 | 1102 | 9055700 |