Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Lxn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016753.4 | NP_058033.2 | 5 | 729 | ||
GenBank mRNA | AK032170.1 | - | 5 | 730 | ||
AK149981.1 | - | 5 | 689 | |||
AK187030.1 | - | 2 | 105 | |||
AK198791.1 | - | 2 | 105 | |||
BC115521.1 | - | 5 | 725 | 40101205 | ||
BC115522.1 | - | 5 | 727 | 40101207 | ||
D88769.1 | - | 5 | 635 |