Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HSPA5 SIRNA Assays ›
          Gene Symbol:
          HSPA5
          Gene Name:
          heat shock protein family A (Hsp70) member 5
          Gene Aliases:
          BIP, GRP78, HEL-S-89n, MIF2
          Chromosome Location:
          Chr.9: 125234848 - 125241387 on Build GRCh38
          UniGene ID:
          Hs.743241
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s6979

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HSPA5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005347.4 NP_005338.1 6 1282
          GenBank mRNA AF188611.1 - 6 967
          AF216292.1 - 6 1021
          AJ271729.1 - 6 1034
          AK129617.1 - 2 66
          AK293604.1 - 7 1108
          BC020235.1 - 6 1225 5020098
          EU794617.1 - 6 1034
          X87949.1 - 6 1240

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          ER overload response
          activation of signaling protein activity involved in unfolded protein response
          cerebellum structural organization
          cerebellar Purkinje cell layer development
          substantia nigra development
          positive regulation of cell migration
          ER-associated ubiquitin-dependent protein catabolic process
          negative regulation of transforming growth factor beta receptor signaling pathway
          endoplasmic reticulum unfolded protein response
          positive regulation of protein ubiquitination
          protein folding in endoplasmic reticulum
          maintenance of protein localization in endoplasmic reticulum
          IRE1-mediated unfolded protein response
          PERK-mediated unfolded protein response
          ATF6-mediated unfolded protein response
          cellular response to glucose starvation
          negative regulation of apoptotic process
          regulation of protein folding in endoplasmic reticulum
          cellular response to antibiotic
          cellular response to manganese ion
          cellular response to interleukin-4
          negative regulation of protein homodimerization activity
          cell-cell adhesion
          toxin transport
          regulation of ATF6-mediated unfolded protein response
          regulation of IRE1-mediated unfolded protein response
          regulation of PERK-mediated unfolded protein response
          positive regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
          glycoprotein binding
          calcium ion binding
          protein binding
          ATP binding
          ATPase activity
          enzyme binding
          protein domain specific binding
          ubiquitin protein ligase binding
          ribosome binding
          unfolded protein binding
          chaperone binding
          misfolded protein binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-7wptj:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0