Search
Search

Mgat3| Gene Symbol: Mgat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010795.4 | NP_034925.2 | 2 | 1119 | ||
| GenBank mRNA | AK081850.1 | - | 2 | 1186 | ||
| BC053040.1 | - | 2 | 1029 | 6406273 | ||
| BC059828.1 | - | 1 | 1011 | 5706186 | ||