Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ARHGDIA SIRNA Assays ›
          Gene Symbol:
          ARHGDIA
          Gene Name:
          Rho GDP dissociation inhibitor alpha
          Gene Aliases:
          GDIA1, HEL-S-47e, NPHS8, RHOGDI, RHOGDI-1
          Chromosome Location:
          Chr.17: 81867719 - 81871406 on Build GRCh38
          UniGene ID:
          Hs.159161
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s699
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ARHGDIA
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001185077.2 NP_001172006.1 6 1481
          NM_001185078.2 NP_001172007.1 6 1119
          NM_001301240.1 NP_001288169.1 7 847
          NM_001301241.1 NP_001288170.1 7 1077
          NM_001301242.1 NP_001288171.1 7 1139
          NM_001301243.1 NP_001288172.1 5 1386
          NM_004309.5 NP_004300.1 6 1251
          NR_125441.1 5 1174
          XM_011523574.1 XP_011521876.1 5 1611
          GenBank mRNA AK293147.1 - 7 1117
          AK300816.1 - 6 1092
          BC005851.2 - 6 1207 2961556
          BC005875.2 - 6 1207 2961556
          BC007630.1 - 1 248
          BC008701.3 - 5 1142 2966760
          BC009759.2 - 6 1172 3935956
          BC013846.1 - 6 1016
          BC016031.1 - 5 1123 3911525
          BC016185.1 - 6 1167 4867857
          BC024258.1 - 6 1167 3027721
          BC027730.2 - 6 1170 4764249
          BC028333.1 - 7 1092
          BC075827.1 - 6 1167 6074977
          BC106044.1 - 6 1210 6162284
          BF343803.1 - 306
          EU794615.1 - 5 1142
          X69550.1 - 6 1168

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00366347_g1

          Panther Classification:

          Molecular Function -

          G-protein modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          movement of cell or subcellular component
          negative regulation of cell adhesion
          regulation of Rho protein signal transduction
          negative regulation of apoptotic process
          positive regulation of GTPase activity
          negative regulation of axonogenesis
          positive regulation of axonogenesis
          regulation of small GTPase mediated signal transduction
          semaphorin-plexin signaling pathway
          regulation of actin cytoskeleton reorganization
          Rho GDP-dissociation inhibitor activity
          GTPase activator activity
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0