Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NGFR SIRNA Assays ›
          Gene Symbol:
          Ngfr
          Gene Name:
          nerve growth factor receptor (TNFR superfamily, member 16)
          Gene Aliases:
          LNGFR, Tnfrsf16, p75, p75NGFR, p75NTR
          Chromosome Location:
          Chr.11: 95568818 - 95587698 on Build GRCm38
          UniGene ID:
          Mm.283893
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s70587

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ngfr
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_033217.3 NP_150086.2 4 900
          GenBank mRNA AF105292.1 - 4 758
          AK037248.1 - 4 901
          BC038365.1 - 4 891 5367638

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          apoptotic process
          signal transduction
          multicellular organism development
          nervous system development
          axon guidance
          central nervous system development
          response to wounding
          regulation of gene expression
          positive regulation of cell death
          negative regulation of neuron projection development
          programmed cell death
          detection of temperature stimulus
          sensory perception of pain
          nerve development
          cell differentiation
          hair follicle morphogenesis
          positive regulation of myelination
          positive regulation of synaptic transmission, cholinergic
          circadian regulation of gene expression
          cellular response to oxidative stress
          positive regulation of Rho protein signal transduction
          negative regulation of fibroblast growth factor receptor signaling pathway
          positive regulation of odontogenesis of dentin-containing tooth
          glucose homeostasis
          positive regulation of apoptotic process
          positive regulation of MAPK cascade
          negative regulation of neuron apoptotic process
          skin development
          positive regulation of fibroblast proliferation
          rhythmic process
          negative regulation of hair follicle development
          positive regulation of protein kinase B signaling
          negative regulation of mitochondrial depolarization
          positive regulation of synaptic transmission, glutamatergic
          negative regulation of cell death
          positive regulation of neuron death
          regulation of reactive oxygen species metabolic process
          positive regulation of apoptotic signaling pathway
          regulation of glucose import in response to insulin stimulus
          beta-amyloid binding
          death receptor activity
          neurotrophin TRKA receptor binding
          protein binding
          Rab GTPase binding
          ubiquitin protein ligase binding
          neurotrophin binding
          nerve growth factor binding
          preprotein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0