Search
Search

Nid1| Gene Symbol: Nid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010917.2 | NP_035047.2 | 15 | 3140 | ||
| GenBank mRNA | AK084876.1 | - | 15 | 3079 | ||
| AK144878.1 | - | 15 | 3105 | |||
| AK166779.1 | - | 15 | 3076 | |||
| BC052939.1 | - | 15 | 3135 | |||
| BC131669.1 | - | 15 | 3051 | 40110859 | ||
| X14194.1 | - | 15 | 3051 | |||
| X14480.1 | - | 15 | 3061 | |||