Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NKX2-5 SIRNA Assays ›
          Gene Symbol:
          Nkx2-5
          Gene Name:
          NK2 homeobox 5
          Gene Aliases:
          Csx, Nkx-2.5, Nkx2.5, tinman
          Chromosome Location:
          Chr.17: 26838664 - 26844994 on Build GRCm38
          UniGene ID:
          Mm.41974
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s70620

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Nkx2-5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_008700.2 NP_032726.1 2 1474
          GenBank mRNA AK142146.1 - 2 1379
          X75415.1 - 2 1538

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          homeodomain transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          vasculogenesis
          heart looping
          heart morphogenesis
          outflow tract septum morphogenesis
          outflow tract morphogenesis
          cardiac conduction system development
          atrioventricular node development
          bundle of His development
          Purkinje myocyte differentiation
          cardiac ventricle morphogenesis
          cardiac ventricle formation
          right ventricular cardiac muscle tissue morphogenesis
          ventricular trabecula myocardium morphogenesis
          apoptotic process involved in heart morphogenesis
          septum secundum development
          proepicardium development
          pulmonary myocardium development
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          multicellular organism development
          endoderm development
          heart development
          adult heart development
          positive regulation of cell proliferation
          positive regulation of gene expression
          negative regulation of cardiac muscle cell apoptotic process
          positive regulation of transcription via serum response element binding
          positive regulation of sodium ion transport
          negative regulation of myotube differentiation
          hemopoiesis
          cell differentiation
          BMP signaling pathway
          thyroid gland development
          embryonic heart tube development
          negative regulation of apoptotic process
          tongue development
          sarcomere organization
          positive regulation of neuron differentiation
          positive regulation of heart contraction
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          spleen development
          cardiac muscle tissue development
          positive regulation of cardioblast differentiation
          ventricular cardiac myofibril assembly
          cardiac muscle cell differentiation
          cardiac muscle tissue morphogenesis
          atrial cardiac muscle cell development
          ventricular cardiac muscle cell development
          regulation of cardiac muscle contraction
          pharyngeal system development
          cardiac muscle cell proliferation
          regulation of cardiac muscle cell proliferation
          heart contraction
          cardiac muscle contraction
          canonical Wnt signaling pathway
          positive regulation of transcription initiation from RNA polymerase II promoter
          heart trabecula formation
          ventricular septum morphogenesis
          atrial septum morphogenesis
          atrioventricular node cell development
          atrioventricular node cell fate commitment
          embryonic heart tube left/right pattern formation
          cardiovascular system development
          negative regulation of canonical Wnt signaling pathway
          positive regulation of voltage-gated calcium channel activity
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II transcription cofactor activity
          transcriptional activator activity, RNA polymerase II transcription factor binding
          DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
          protein binding
          transcription factor binding
          serum response element binding
          protein homodimerization activity
          histone deacetylase binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0