Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Orm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013623.2 | NP_038651.2 | 5, 6 | 589 | ||
GenBank mRNA | BC141235.1 | - | 5, 6 | 593 | 9055847 | |
BC145559.1 | - | 5, 6 | 593 | 9054123 | ||
BC145560.1 | - | 5, 6 | 623 | |||
BC171932.1 | - | 4, 5 | 450 |