Search
Search

Pdgfb| Gene Symbol: Pdgfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011057.3 | NP_035187.2 | 4 | 1117 | ||
| GenBank mRNA | AF162784.1 | - | 4 | 307 | ||
| BC023427.1 | - | 3 | 241 | |||
| BC053430.1 | - | 4 | 1117 | 6330609 | ||
| BC064056.1 | - | 4 | 784 | 1380782 | ||