Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Abcb1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011076.2 | NP_035206.2 | 21 | 2670 | ||
GenBank mRNA | AK144907.1 | - | 3 | 399 | ||
AY864315.1 | - | 20 | 2533 | |||
M24417.1 | - | 15 | 2019 | |||
M30697.1 | - | 21 | 2684 | |||
M33581.1 | - | 21 | 2669 |