Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ppp3ca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293622.1 | NP_001280551.1 | 2 | 968 | ||
NM_008913.5 | NP_032939.1 | 2 | 968 | |||
GenBank mRNA | AK146387.1 | - | 2 | 912 | ||
AK150393.1 | - | 2 | 327 | |||
AK166287.1 | - | 2 | 745 | |||
AK209023.1 | - | 1 | 109 | |||
BC096652.1 | - | 2 | 210 | |||
BC138612.1 | - | 2 | 241 | 8861632 | ||
J05479.1 | - | 2 | 265 |