Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Psme2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029855.1 | NP_001025026.1 | 7 | 433 | ||
NM_011190.3 | NP_035320.1 | 7 | 533 | |||
GenBank mRNA | AA267668.1 | - | 7 | 424 | ||
AK012344.1 | - | 7 | 462 | |||
AK088654.1 | - | 7 | 460 | |||
AK143556.1 | - | 7 | 465 | |||
BC005680.1 | - | 7 | 453 | 3154255 | ||
BC057859.1 | - | 7 | 530 | 5310526 | ||
BU563130.1 | - | 7 | 602 | |||
CA536627.1 | - | 7 | 445 | |||
U60329.1 | - | 7 | 402 |
Gene Symbol: Psme2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281472.1 | NP_001268401.1 | 1 | 693 | ||
GenBank mRNA | D87910.1 | - | 1 | 420 |