Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PTK2B SIRNA Assays ›
          Gene Symbol:
          Ptk2b
          Gene Name:
          PTK2 protein tyrosine kinase 2 beta
          Gene Aliases:
          CADTK, CAKB, CAKbeta, E430023O05Rik, FADK2, FAK2, PYK2, Raftk
          Chromosome Location:
          Chr.14: 66153257 - 66281066 on Build GRCm38
          UniGene ID:
          Mm.21613
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s72405

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ptk2b
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001162365.1 NP_001155837.1 18, 19 1858
          NM_001162366.1 NP_001155838.1 17, 18 1688
          NM_172498.3 NP_766086.2 18, 19 1858
          GenBank mRNA AK088696.1 - 18, 19 1859
          AK150105.1 - 18, 19 1789
          BC137704.1 - 17, 18 1683 8860723
          BC144849.1 - 17, 18 1683 9053397
          JN971017.1 - 17, 18 1651

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_017318, XM_006252145, XM_006252143, XM_008770774, XM_006252144, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          angiogenesis
          oocyte maturation
          positive regulation of cell-matrix adhesion
          sprouting angiogenesis
          marginal zone B cell differentiation
          immune system process
          protein phosphorylation
          cellular defense response
          cytoskeleton organization
          actin filament organization
          cell adhesion
          cell surface receptor signaling pathway
          signal complex assembly
          epidermal growth factor receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          integrin-mediated signaling pathway
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          regulation of cell shape
          positive regulation of endothelial cell migration
          negative regulation of muscle cell apoptotic process
          regulation of cGMP-mediated signaling
          regulation of macrophage chemotaxis
          positive regulation of neuron projection development
          phosphorylation
          cell migration
          peptidyl-tyrosine phosphorylation
          regulation of cell adhesion
          negative regulation of ossification
          positive regulation of cell growth
          positive regulation of cell migration
          negative regulation of bone mineralization
          regulation of cGMP biosynthetic process
          positive regulation of actin filament polymerization
          neuron projection development
          regulation of inositol trisphosphate biosynthetic process
          tumor necrosis factor-mediated signaling pathway
          ionotropic glutamate receptor signaling pathway
          peptidyl-tyrosine autophosphorylation
          response to hydrogen peroxide
          activation of Janus kinase activity
          negative regulation of apoptotic process
          stress fiber assembly
          negative regulation of potassium ion transport
          positive regulation of JUN kinase activity
          negative regulation of neuron apoptotic process
          blood vessel endothelial cell migration
          positive regulation of phosphatidylinositol 3-kinase activity
          innate immune response
          regulation of nitric oxide biosynthetic process
          bone resorption
          negative regulation of myeloid cell differentiation
          positive regulation of translation
          positive regulation of angiogenesis
          positive regulation of protein kinase activity
          positive regulation of JNK cascade
          vascular endothelial growth factor receptor signaling pathway
          focal adhesion assembly
          positive regulation of peptidyl-tyrosine phosphorylation
          regulation of calcium-mediated signaling
          positive regulation of nitric-oxide synthase activity
          regulation of release of sequestered calcium ion into cytosol
          positive regulation of synaptic transmission, glutamatergic
          long-term synaptic potentiation
          chemokine-mediated signaling pathway
          positive regulation of ERK1 and ERK2 cascade
          cellular response to retinoic acid
          cellular response to fluid shear stress
          activation of GTPase activity
          regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          regulation of establishment of cell polarity
          regulation of actin cytoskeleton reorganization
          regulation of N-methyl-D-aspartate selective glutamate receptor activity
          positive regulation of reactive oxygen species metabolic process
          positive regulation of excitatory postsynaptic potential
          positive regulation of B cell chemotaxis
          nucleotide binding
          protein kinase activity
          calmodulin-dependent protein kinase activity
          protein tyrosine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          signal transducer activity
          NMDA glutamate receptor activity
          receptor binding
          protein binding
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          protein complex binding
          3-phosphoinositide-dependent protein kinase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-v2dhc:80/100.66.79.56:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0