Search
Search

Ptpn11| Gene Symbol: Ptpn11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109992.1 | NP_001103462.1 | 13 | 1715 | ||
| NM_011202.3 | NP_035332.1 | 13 | 1727 | |||
| GenBank mRNA | AK147354.1 | - | 12 | 3561 | ||
| AK159501.1 | - | 13 | 1715 | |||
| AK159587.1 | - | 13 | 1715 | |||
| AK163809.1 | - | 10 | 1143 | |||
| BC057398.1 | - | 13 | 1659 | 6409640 | ||
| BC059278.1 | - | 13 | 1693 | 2866038 | ||
| D84372.1 | - | 13 | 1603 | |||