Search
Search

Ptpn11| Gene Symbol: Ptpn11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109992.1 | NP_001103462.1 | 10 | 1378 | ||
| NM_011202.3 | NP_035332.1 | 10 | 1378 | |||
| GenBank mRNA | AK147354.1 | - | 9 | 3224 | ||
| AK159501.1 | - | 10 | 1378 | |||
| AK159587.1 | - | 10 | 1378 | |||
| AK163809.1 | - | 7 | 806 | |||
| BC057398.1 | - | 10 | 1310 | 6409640 | ||
| BC059278.1 | - | 10 | 1356 | 2866038 | ||
| D84372.1 | - | 10 | 1254 | |||
| S59187.1 | - | 3 | 296 | |||