Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rab5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB232595.1 | - | 2 | 206 | ||
AK089199.1 | - | 3 | 408 | |||
AK134691.1 | - | 3 | 347 | |||
AK159832.1 | - | 3 | 397 | |||
AK167599.1 | - | 3 | 397 | |||
AK170540.1 | - | 3 | 417 | |||
AK172328.1 | - | 3 | 354 | |||
BC023027.1 | - | 3 | 363 | 5369566 | ||
BC027378.1 | - | 2 | 169 | |||
BC029678.1 | - | 3 | 347 | 5368061 | ||
M79312.1 | - | 2 | 110 |