Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RARG SIRNA Assays ›
          Gene Symbol:
          Rarg
          Gene Name:
          retinoic acid receptor, gamma
          Gene Aliases:
          Nr1b3, RAR-gamma, RARD, RARgamma2
          Chromosome Location:
          Chr.15: 102234938 - 102257522 on Build GRCm38
          UniGene ID:
          Mm.1273
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s72736

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rarg
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001042727.2 NP_001036192.1 4 801
          NM_011244.4 NP_035374.3 6 1039
          GenBank mRNA AK148064.1 - 6 1036
          AK155911.1 - 4 654
          BC012923.1 - 6 965 3154037
          M34475.1 - 4 577
          M34476.1 - 6 796
          X15848.1 - 6 859

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          C4 zinc finger nuclear receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          neural tube closure
          chondrocyte development
          glandular epithelial cell development
          retinal pigment epithelium development
          growth plate cartilage development
          growth plate cartilage chondrocyte growth
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          anterior/posterior pattern specification
          regulation of gene expression
          positive regulation of gene expression
          embryonic camera-type eye development
          regulation of myelination
          negative regulation of chondrocyte differentiation
          response to retinoic acid
          embryonic hindlimb morphogenesis
          multicellular organism growth
          camera-type eye development
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          positive regulation of programmed cell death
          steroid hormone mediated signaling pathway
          negative regulation of cell differentiation
          regulation of myeloid cell differentiation
          positive regulation of transcription from RNA polymerase II promoter
          embryonic eye morphogenesis
          retinoic acid receptor signaling pathway
          reproductive structure development
          gland development
          retina development in camera-type eye
          limb development
          face development
          bone development
          bone morphogenesis
          epithelium development
          trachea cartilage development
          prostate gland epithelium morphogenesis
          negative regulation of cartilage development
          Harderian gland development
          cellular response to retinoic acid
          RNA polymerase II regulatory region sequence-specific DNA binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          steroid hormone receptor activity
          retinoic acid receptor activity
          protein binding
          zinc ion binding
          sequence-specific DNA binding
          metal ion binding
          retinoid X receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0